Learn More
Medchemexpress LLC EXP3179 100mg | 114798-36-6 | 100 MG

Supplier: Medchemexpress LLC HY114950100MG
EXP3179 (Losartan carboxaldehyde) is an aldehyde metabolite and synthetic intermediate of losartan supplied for research use. It is provided as a solid for biochemical and pharmacological research, with manufacturer guidance for storage and formulation.
- Intended for research use only, not for human or clinical use.
- Molecular formula C22H21ClN6O; molecular weight 420.89.
- Soluble in DMSO (≈125 mg/mL); in vivo formulation examples provided by manufacturer.
- Recommended storage: powder at -20°C; in solution -80°C for 6 months or -20°C for 1 month.
- Supplied in a 100 mg quantity for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.