Learn More
Medchemexpress LLC EXP3179 50mg | 114798-36-6 | 420.89 g/mol | C22H21ClN6O | 50 MG

Supplier: Medchemexpress LLC HY11495050MG
EXP3179 (losartan carboxaldehyde) is an aldehyde metabolite of losartan that inhibits endothelial cyclooxygenase-2 (COX-2). Supplied as a white to off-white solid for research use, it is intended for studies of inflammation and vascular biology.
- High purity (98.09%).
- Molecular formula C22H21ClN6O; molecular weight 420.89 g/mol.
- Supplied as 50 mg powder.
- Appearance: white to off-white solid.
- Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.