Learn More
MSC Sealed Air Instapak Quick RT Bags, 18x24 in, 30/Case
Supplier:  MSC 39590443
Encompass
Instapak Quick® RT expandable foam bags provide on-demand product protection without requiring specialized equipment. These bags expand up to 27 times their original size in seconds, forming custom-fit protective foam cushions ideal for low-volume packaging operations.
  • Expands on demand, optimizing storage space
  • Activates with simple press, pat, and pack technique
  • Provides custom-fit, heavy-duty cushioning, bracing, and light blocking
  • Requires no preheating warming unit or dispensing system
  • Chemicals activate in the bag, eliminating cleanup
  • Recommended temperature range of 65° - 90° F
Catalog No. 50-191-9429