Learn More
Enzo Life Sciences Express DNA methylation kit (200tests)
SDP
Supplier:  Enzo Life Sciences ENZ450010200
Encompass_Preferred
The Express DNA methylation kit can be used for rapid and reliable bisulfite treatment and conversion of DNA for methylation analysis. Simply add the Express Conversion Reagent for the reaction to occur. With the addition of heat, DNA denaturation is facilitated. Desulphonation and clean-up of the converted DNA is performed using a unique low-elution spin column. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing, etc.
Catalog No. 50-201-2562