Learn More
Cole-Parmer Extech EX350 TrueRMS Multimeter w/ Noncontact Voltage Detection
Supplier:  Cole-Parmer 2002701
Encompass
Increase safety of technicians with noncontact voltage detection and LoZ mode. True RMS and low pass filter provide accurate readings of variable frequency signals.
Catalog No. NC1518503