Learn More
Extech Instrument Corporation Extech IR200: Non-Contact Forehead InfraRed Thermometer
Supplier:  Extech Instrument Corporation IR200
Encompass_Preferred
Due to high demand and limited supply, once a PO has been placed the order cannot be cancelled and is non-returnable. This product has an extremely volatile lead time and availability. Expected ship date can change from original promise date.
Ideal for quickly screening individuals for elevated skin temp. This infrared thermometer has a non-contact design that eliminates the need for replacement probe covers and other supplies. The IR200 is accurate to 0.5°F with 0.1°F/°C resolution when measuring skin temp. between 89.6°F to 108.5°F. Adjustable alarm alerts user visually and audibly when temp. exceeds programmed limit. Optimum measurement distance of 1.9 in. to 5.9 in. Move the Body/Surface function switch to the Surface position for non-contact surface temp. measurements from 32°F to 140°F. Fast response of just 0.5 seconds and large backlit LCD display. Memory stores up to 32 readings and comes complete with 2 AA (1.5V) batteries and pouch.
Catalog No. NC0781598
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.