Learn More
Quantum Storage Systems EXTENDED LABEL HOLDER PK24
Supplier:  Quantum Storage Systems ELH410
Encompass_Preferred
Optional extended label holders are designed for large bar code sanning. Label holders fit all Shelf Bins.
Catalog No. 50-752-0095