Learn More
Grainger Extension Cord 50ft 12Ga 20A SOW Blk
Supplier:  Grainger 14N240
Encompass_Preferred
The Grainger 14N240 is a 14 gauge, type N thermocouple wire with fiberglass insulation and a stainless steel overbraid. It has a temperature range of -454 to 2282°F (-270 to 1250°C). The wire has nickel alloy conductors and can be used with extension or thermocouple wire with the same specifications. It is 100 feet in length and 1/8 inch in diameter. The wire is flexible, moisture and abrasion resistant. It can be used in various temperature measurement applications.
Catalog No. 50-274-8919