Learn More
Ace Glass, Inc. Extension lead, K thermocouple, 96in, PTFE covered, male and female mini plug ends
SDP
Supplier:  Ace Glass, Inc. 1211328
Encompass
96 PTFE-covered extension lead for temperature probes, has a type K thermocouple, and male and female mini plug ends. Extension lead allows the use of a temperature controller up to 8 away.
Catalog No. 50-977-462