Learn More
Med Vet International External Fixation Rod, J0371RMA, 3/16" x 2", Each
Supplier:  Med Vet International JOR371RMA
Encompass_Preferred
Carbon fiber ESF bars offer two advantages over conventional stainless. They are radiolucent and lighter in weight but comparable in strength. Note carbon fiber need to cut with a saw versus stainless pin cutters.
Catalog No. 50-005-81927