Learn More
Grainger Externalooving Blade KGDL 2T17C Ext
Supplier:  Grainger 170HV6
Encompass_Preferred
The Grainger External Grooving Blade KGDL-2T17C-170HV6 is a high-speed steel blade with a width of 0.078 inches and a thickness of 0.078 inches. It has a 170 degree included angle and is designed for external grooving, turning, and cutoff applications. The blade has a TiN coating for increased hardness and lubricity. It is made in the USA and fits standard tool holders with a 0.312 inch shank diameter. The overall length is 3 inches. This durable blade is ideal for use on steel, cast iron, aluminum, and other materials.
Catalog No. 50-276-4752