Learn More
Grainger Externalooving Blade KGDR 5T10C Ext
Supplier:  Grainger 170HW9
Encompass_Preferred
The 170HW9 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor manufactured by Baldor-Reliance. It has a 1.15 service factor, CCW rotation when facing the shaft end, and features a cast iron frame, cast iron junction box, and cast iron end brackets. The motor is totally enclosed, fan cooled (TEFC) with a 1" diameter shaft and measures approximately 9-1/4" long x 7-3/4" wide x 8-7/8" high.
Catalog No. 50-276-4357