Learn More
Grainger Externalooving Toolholder KGBAL 163
Supplier:  Grainger 170JA5
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JA5 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and accepts grooving inserts with thicknesses ranging from 0.078 to 0.197 inches. It is made from forged steel for durability and has a black oxide finish to prevent corrosion. The 170JA5 is compatible with ISO standard grooving inserts and provides rigid clamping to minimize insert movement during machining. This external grooving toolholder enables precise grooving in a variety of materials.
Catalog No. 50-276-4420