Learn More
Grainger Externalooving Toolholder KGDL 1212J
Supplier:  Grainger 170JW8
Encompass_Preferred
The Walter Indexable Parting and Grooving Insert Holder 56NP14 is a tool holder for parting and grooving inserts. It has a nominal shank diameter of 1 inch, an overall length of 4 inches, and accepts inserts with a thickness of 0.118 inches. The holder is made from steel and has a black oxide finish. It can be used for parting, grooving, and threading operations on lathes, screw machines, and CNC machines. The holder accepts negative rake inserts with a .118 inch thickness to provide rigidity and precision during parting and grooving operations.
Catalog No. 50-276-4166