Learn More
Grainger Externalooving Toolholder KGDL 2020K
Supplier:  Grainger 170JL4
Encompass_Preferred
The 170JL4 is a Dayton 1/3 HP, 115/230V, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Electric Motor. It has a 1/3 horsepower, operates on 115/230 volt single phase power, has a 1725 RPM at 60 Hz, a 56C frame size, open drip proof enclosure, and ball bearings. The motor is thermally protected, has manual reset, and is CSA certified. It can be used for various light industrial applications requiring 1/3 HP.
Catalog No. 50-276-4656