Learn More
Grainger Externalooving Toolholder KGDR 16C
Supplier:  Grainger 170JK8
Encompass_Preferred
The Kyocera External Grooving Toolholder 61NR16 is a toolholder for external grooving and parting. It has a right hand orientation and accepts inserts with a .157" thickness. The toolholder body is made from steel with a black oxide finish and has a 1/2" shank diameter. It is suitable for use on most CNC lathes and manual lathes. The toolholder allows for high precision grooving, parting and cutoff operations. It has a maximum insert width capacity of .472" and a maximum depth of cut capacity of .315". The toolholder requires a screw clamp style insert for secure holding. It is an affordable and durable option for grooving applications.
Catalog No. 50-276-4170