Learn More
Grainger Externalooving Toolholder KGDR 16X4T
Supplier:  Grainger 170JP4
Encompass_Preferred
The Grainger Approved External Grooving Toolholder is a toolholder designed to hold grooving inserts for external grooving operations. It has a shank diameter of 1/2", an overall length of 4", and accepts inserts with a thickness of 3/64" to 1/8". The toolholder is made from steel and has a black oxide finish. It can be used for grooving, cutoff, and threading applications on lathes, automatic lathes, and screw machines. The toolholder accepts standard ANSI style inserts and provides rigid clamping to minimize insert movement.
Catalog No. 50-276-4196