Learn More
Grainger Externalooving Toolholder KGDR 1024J
Supplier:  Grainger 170JV9
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JV9 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and accepts grooving inserts with thicknesses ranging from 0.078 to 0.197 inches. It is made from steel and has an ISO designation of PCLNR/L 2020K12. The toolholder is suitable for use on CNC lathes and machining centers for grooving, parting off, and cutoff applications on materials like steel, stainless steel, cast iron, and non-ferrous metals. It enables precise grooving in locations with limited tool access.
Catalog No. 50-276-4304