Learn More
Grainger Externalooving Toolholder KGDR 2525M
Supplier:  Grainger 170JM9
Encompass_Preferred
The Kyocera External Grooving Toolholder 61NR08 is a toolholder designed for external grooving operations. It has a shank diameter of 1 inch, an overall length of 4 inches, and accepts inserts with a thickness of 0.078-0.118 inches. The toolholder is made of steel and has a black oxide finish. It features a screw clamp to hold inserts in place. The 61NR08 is suitable for use on most lathes, machining centers, and other machine tools. It enables the machining of external grooves, parting, cutoff operations and more when paired with the appropriate insert. This toolholder requires a minimum bore diameter of 1.06 inches.
Catalog No. 50-276-3819