Learn More
Grainger Externalooving Toolholder KGDR 164T2
Supplier:  Grainger 170JK0
Encompass_Preferred
The Kyocera KGDR 164T20 is an external grooving toolholder with a 20mm shank diameter and 164mm overall length. It has a T-style clamp and is made of steel for rigidity and durability. The toolholder accepts ISO turning inserts with a .078" thickness, allowing for a wide range of insert geometries. It can be used for external grooving, parting, and cutoff applications on most common materials. The toolholder body has a black oxide finish for corrosion resistance. Overall, the KGDR 164T20 provides a rigid and precise platform for external grooving operations.
Catalog No. 50-276-4269