Learn More
Grainger Externalooving Toolholder KGDR 16X4T
Supplier:  Grainger 170JP8
Encompass_Preferred
The 170JP8 is a 3M Scotch-Brite General Purpose Hand Pad, 8" x 4" x 1/2", made of high-quality nylon fibers in an open textured nonwoven construction. This durable, long-lasting pad has an abrasive on all sides for scrubbing and cleaning. It effectively removes grease, oil, dirt, carbon, food residue, and more from a variety of surfaces including stainless steel, aluminum, fiberglass, and painted metal. The pad has an open web design that rinses easily. For heavy duty scrubbing.
Catalog No. 50-276-4445