Learn More
Grainger Externalooving Toolholder KGDR 12X3T
Supplier:  Grainger 170JT6
Encompass_Preferred
The 170JT6 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Electric Motor. It has a 1/3 horsepower, operates on 115/230 volt AC power, has a speed of 1725 RPM, uses a 56C frame, is open drip proof, utilizes ball bearings, and is designed for general purpose applications. The motor is thermally protected, suitable for continuous duty applications, and can be mounted horizontally or vertically. It measures approximately 9-1/2" long x 5-3/4" wide x 6-7/8" high.
Catalog No. 50-276-4660