Learn More
Grainger Externalooving Toolholder KGDR 2020K
Supplier:  Grainger 170JM4
Encompass_Preferred
The Kyocera External Grooving Toolholder is a high-quality toolholder designed for external grooving operations. It features a precision-ground body made from SCM435 steel for durability. The toolholder has a ISO S-DKB style shank with a width of 25mm. It accepts ISO SCLCR/L 06-25 style inserts with a thickness of 2.5mm. The toolholder provides rigid clamping and precise insert positioning for accurate grooving. With its durable construction and ability to hold precision inserts, this toolholder is well-suited for demanding external grooving applications.
Catalog No. 50-276-4241