Learn More
Grainger Externalooving Toolholder KGDR 1616H
Supplier:  Grainger 170JM1
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JM1 is designed for external grooving operations. It features a JM style interface and accepts ISO turning inserts up to 16mm. Made from steel, it supports both right and left hand turning. With a length of 4 inches and a shank diameter of 1 inch, it offers a cutting depth range of 0.039 - 0.197 inches. Ideal for CNC lathes, it handles external grooving, parting, and cutoff on materials like steel, stainless steel, cast iron, and non-ferrous metals. This durable toolholder ensures rigid clamping for accurate grooving.
Catalog No. 50-276-4242