Learn More
Grainger Externalooving Toolholder KKCL 122B
Supplier:  Grainger 170HX2
Encompass_Preferred
The External Grooving Toolholder KKCL-122B-170HX2 by Kennametal is a right-hand external grooving toolholder with a 1/2" shank diameter and a maximum groove width of 0.122". It has an American National Standard Code (ANSI) insert style of 170 with a hook-lever clamp and is made of steel. The toolholder accepts inserts with a thickness of 0.118" to 0.197" and has a maximum depth of cut of 0.315". It can be used for external grooving, profiling and cutoff applications. The toolholder allows for high feed rates and secure clamping of inserts.
Catalog No. 50-276-4734