Learn More
Grainger Externalooving Toolholder KKCL 162C
Supplier:  Grainger 170HX5
Encompass_Preferred
The KYOCERA External Grooving Toolholder KKCL-162C-170HX5 is a toolholder for external grooving and turning. It has a clamping range of 0.157-0.394 inches and accepts inserts with a thickness of 0.118 inches. The toolholder is made of steel, has a right hand orientation, and features a top clamp. It has an overall length of 6.69 inches, a shank height of 1.26 inches, and a shank width of 1.26 inches. The KKCL-162C-170HX5 has an American National Standards Institute (ANSI) style shank, and is suitable for use on CNC lathes.
Catalog No. 50-276-4793