Learn More
Grainger Externalooving Toolholder KKCR 164D
Supplier:  Grainger 170HZ6
Encompass_Preferred
The Kyocera External Grooving Toolholder 170HZ6 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and accepts grooving inserts with a thickness of 0.118 inches. It is made of steel and is black oxide coated. The 170HZ6 has an American National Standards Institute (ANSI) style toolholder block to accommodate standard inserts. This toolholder enables rigid setups for precise grooving in turning applications.
Catalog No. 50-276-4779