Learn More
Fine Science Tools Extra Fine Graefe Forceps, 10cm
SDP
Supplier:  Fine Science Tools 1115510
Encompass
Tip Shape: Curved/ Tips: 1x2 Teeth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9020845