Learn More
IW Tremont Co Inc LabExact Extraction thimbles Cellulose Format: 60x180 mm (Inner diameter x Height) pack of 25
SDP
Supplier:  IW Tremont Co Inc LECT5039
Encompass_Preferred
LabExact High quality extraction thimbles available in a variety of dimensions to fit most Soxhlet extraction units. Extraction thimbles are manufactured with high purity cellulose fibers and have a consistent 2.0mm wall thickness. Common applications include air and waste gas analysis, collection of solid particles such as dust, concrete slurry evaluation, analysis of pesticide residues, oil/fat content of solid foods and oil and grease analysis of solid wastes.
Catalog No. 50-223-7658