Learn More
Waters Corp Waters Seal Removal Tool, Plastic End, HPLC System
Supplier:  Waters Corp WAT039803
Encompass_Preferred
The Seal Removal Tool is designed to safely remove seals from various pump heads, specifically within HPLC systems. It features both metal and plastic ends, ensuring that plunger seals can be extracted without scratching or damaging the sealing surfaces, unlike conventional tools.
  • Designed to remove plunger seals from pump heads.
  • Features a plastic end to prevent scratching of sealing surfaces.
  • Compatible with O-ring energized and spring-energized seals for various pump types.
  • Equipped with both metal and plastic screw ends for versatile seal removal procedures.
  • Prevents damage to pump head surfaces during seal extraction.
  • Used with HPLC systems.
Catalog No. 50-818-639