Learn More
MSC Bosch Dust Extractor, 14.5 Gallon Capacity, Auto Filter Clean, HEPA Filter
Supplier:  MSC 47824214
Encompass
This 14.5-gallon dust extractor is designed for OSHA silica dust regulation compliance, offering strong airflow and suction. Its automatic filter-cleaning system activates every 15 seconds to maintain maximum suction. Features include power tool activation, allowing a connected tool to control the vacuum, and a Power Broker dial to adjust suction force based on the application.
  • Automatic filter cleaning for continuous maximum suction.
  • HEPA filter captures 99.97% of particles at 0.3 microns or larger.
  • Power tool activation for integrated tool control.
  • Wet vacuuming water-level sensor protects the motor.
  • Rugged construction with a polypropylene tank for durability.
Catalog No. 50-283-8121