Learn More
ARMOR GUYS Extraflex Gloves, 2XL
Supplier:  ARMOR GUYS 043022X
Encompass_Preferred
Range: Extraflex | Liner: 13g, Blue Engineered Yarn Liner | Coating: Black Nitrile Palm Coating With Smooth Finish | Attributes: MicroFoam Coating Ensures Excellent Oil Grip, OLYMPUS GRIP, Reinforced Thumb Crotch | Applications: Bottling, Glass Handling, Heavy Material Handling
Catalog No. NC1592045