Learn More
MSC Clarke Hand Tool, General Purpose, for Carpet Extractors, 4 in Overall Width
Supplier:  MSC 35712926
Encompass
This general purpose hand tool is an accessory designed for use with Clarke CleanTrack carpet extractors. It provides efficient cleaning for various applications, making it suitable for professional use.
  • Accessory type: Hand tool
  • For use with CleanTrack S16, L18, EX40 16ST & 18LX carpet extractors
  • Overall width: 4 in
  • Suitable for high-speed cleaning applications
  • Aids in effective carpet extraction
Catalog No. 50-283-9183