Learn More
Wilmad Labglass EXTRTN APP SXHLT 1.5L COMP
Supplier:  Wilmad Labglass LG6910102
Encompass_Preferred
Extraction Apparatus, Soxhlet, Extra Large, Complete. Extractor ID: 100mm; Extractor Capacity: 1500mL; Condenser Joint Size: [ST] 103/60, Flask Capacity: 3000mL, Flask Joint Size [ST] 29/42
Catalog No. 50-805-412
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.