Learn More
Grainger Eye End Fitting 316 SS 3/32 in Rope dia.
Supplier:  Grainger 16X679
Encompass_Preferred
The Loos Eye End Fitting is a 3/32" compatible eye end fitting made of carbon steel with a zinc finish. It has a working pressure of 3,000 psi and temperature range of -20 to +250°F. This fitting connects 3/32" OD tubing to threaded male NPT ports. It has a hex shaped body for easy tightening with a wrench. The Loos eye end fitting is ideal for use in pneumatic, hydraulic, and other low pressure fluid power systems.
Catalog No. 50-276-5639