Learn More
Grainger Eye End Fitting 316 SS 3/8 in Rope dia.
Supplier:  Grainger 16X687
Encompass_Preferred
The LOOS Eye End Fitting is a 3/8 inch eye end fitting made of forged steel. It has a working pressure of 3000 psi and temperature range of -20 to 300 degrees Fahrenheit. This fitting has a zinc finish and includes a locknut for securing the fitting. It is compatible with 3/8 inch OD rigid metal conduit and is UL listed. The overall dimensions are 1-1/2 inches length by 1 inch width. This eye end fitting provides a secure 90 degree bend in electrical conduit runs.
Catalog No. 50-276-8252