Learn More
Grainger Eye End Fitting SS 1/16 in Rope dia.
Supplier:  Grainger 16X668
Encompass_Preferred
The Loos & Co. Eye End Fitting is a 1/16" NPT eye end fitting made of 316 stainless steel for corrosion resistance. It has a maximum pressure rating of 10,000 psi and temperature rating of up to 800°F. This fitting allows a hose or pipe to be securely connected using the female NPT threads on one end and provides a loop on the other end to attach additional components. The small 1/16" size makes this fitting well-suited for instrumentation, test equipment, and other light duty hydraulic applications. Overall, this stainless steel eye end fitting provides a durable threaded connection point with an attached loop.
Catalog No. 50-276-8253