Learn More
Grainger Eye End Fitting SS 1/8 in Rope dia.
Supplier:  Grainger 16X670
Encompass_Preferred
The Loos & Co. 1/8" Eye End Fitting is a 316 stainless steel fitting with a 1/8" NPT female thread and a welded eye end. It has a maximum pressure rating of 3000 psi at 100°F. The overall length is 1-3/4" and it weighs 0.06 lbs. This fitting allows you to attach threaded pipe to the eye opening for a strong and versatile connection point. It can be used in hydraulic, pneumatic, lubrication, and industrial applications. The 316 stainless steel material offers good corrosion resistance. This eye end fitting is made in the USA by Loos & Co., a company known for high quality hydraulic and pneumatic fittings.
Catalog No. 50-276-7075