Learn More
Grainger Eye End Fitting SS 3/32 in Rope dia.
Supplier:  Grainger 16X669
Encompass_Preferred
The Loos Eye End Fitting is a 3/32 inch compatible eye end fitting made of 316 stainless steel. It has a maximum pressure rating of 5,000 psi and temperature rating of up to 800 degrees Fahrenheit. This fitting has a thread size of 3/32 inch NPT and is manufactured in the United States. It can be used for instrumentation tubing in hydraulic, pneumatic, lubrication, cooling and other systems. This eye end fitting provides a leak-proof seal.
Catalog No. 50-276-5068