Learn More
Grainger Eye End Fitting SS 5/32 in Rope dia.
Supplier:  Grainger 16X671
Encompass_Preferred
The Loos Eye End Fitting is a 5/32" compatible eye end fitting made of carbon steel with a zinc finish. It has a maximum pressure rating of 3000 psi and temperature rating of 300°F. This fitting connects to 5/32" OD rigid or soft copper tubing and is ANSI/NSF 61 approved for potable water. It is made in the USA by Loos & Co. and sold individually. This eye end fitting provides a secure 90 degree connection point for copper tubing in plumbing and HVAC applications.
Catalog No. 50-276-7221