Learn More
Electron Microscopy Sciences Style 5A/45-7, Material: Inox 08. Finish: Matte. OAL: 115 mm. Tip width: 0.13 mm. Tip thickness: 0.08 mm., 1/EA
SDP
Supplier:  Electron Microscopy Sciences 07085A457PS
Encompass_Preferred
Specially designed Dumont eyelash tweezers for the eyelash industry. Due to the demand and nature of the job to be done, we are directing those in the eyelash industry to this tweezer, which is perfect for the task. Fine and oblique tips. Serrated. Fine and oblique tips. Serrated. Material: Inox 08. Finish: Matte . OAL: 115 mm . Tip width: 0.13 mm. Tip thickness: 0.08 mm.
Catalog No. 50-209-0544