Learn More
Zymo Research Corporation EZ-96 DNA Methylation-Lightning™ MagPrep, 4x96 Rxns.
SDP
Supplier:  Zymo Research Corporation D5046
Encompass_Preferred
The EZ-96 DNA Methylation-Lightning MagPrep Kit provides high-throughput and automated methylation analysis. No preparation is necessary with the ready-to-use conversion reagent - simply add this unique reagent to a DNA sample and incubate the reaction for about one hour. Desulphonation and clean-up of the converted DNA is performed while bound to the MagBinding Beads. This kit produces the highest yields of bisulfite-converted DNA that is ideal for PCR, arrays, library preps, and Next-Generation sequencing.
Catalog No. 50-203-7131