Learn More
Apexbio Technology LLC EZ Cap™ mCherry mRNA (5mCTP, UTP), 100ug
SDP
Supplier:  Apexbio Technology LLC R1017.1
Encompass_Preferred
mCherry mRNA encodes the red fluorescent protein, mCherry, which is derived from DsRed, a protein found in Discosoma sea anemones. mCherry is a monomeric fluorophore with a peak absorption at 587 nm and emission at 610 nm. It is stable and resistant to photobleaching.This mRNA is capped using a co-transcriptional capping method, which results in the naturally occuring Cap 1 structure with high capping efficiency. It is polyadenylated, modified with 5mCTP/UTP and optimized for mammalian systems. The addition of 5mCTP/UTP and poly(A) tail can reduce host cell immune response and enhance mRNA stability. It mimics a fully processed mature mRNA. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8494