Learn More
Zymo Research Corporation EZ DNA Methlyation-Direct™ Kit (200 Rxns)
SDP
Supplier:  Zymo Research Corporation D5021
Encompass_Preferred
The EZ DNA Methylation-Direct kit is a bisulfite conversion kit that features reliable and complete DNA bisulfite conversion directly from blood, tissue, and cells without the prerequisite for DNA purification. The increased sensitivity of this bisulfite conversion kit makes it possible to amplify bisulfite-converted DNA from as few as 10 cells or 50 pg DNA. Recovered bisulfite-converted DNA is ideal for downstream analyses including PCR amplification, endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-444-327