Learn More
Foxx Life Sciences Foxx Life Sciences™ Carboy Vent Filters
Supplier:  Foxx Life Sciences 25H1608OEM
Encompass_Preferred
Foxx Life Sciences™ Carboy Vent Filters are used for hydrophobic venting and solvent filtration applications. The PTFE filter membrane is ideal for maintaining purified water stored in carboys, or for sterile venting during a slow exhaust/liquid autoclave cycle of EZgrip carboys.
Catalog No. NC2079934