Learn More
Novus Biologicals EZ Standard Pack 2
SDP
Supplier:  Novus Biologicals PSST02EZ8
Encompass
Compatible with Jess, Abby, Wes, Peggy Sue and Sally Sue systems. Pack includes 8 packets with one each of the Single Use Biotinylated Ladder (MW ladder spans 12-230 kDa) and Single Use FL Standard (180 kDa system control protein) vials. Pack contains enough reagents to obtain 200 Simple Western data points.
Catalog No. NC1602260