Learn More
Medchemexpress LLC Ezatiostat | 168682-53-9 | 99.7% | 529.65 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13634A50MG
Encompass_Preferred
Ezatiostat is a tripeptide analog of glutathione that acts as a selective, orally active glutathione S-transferase P1-1 (GSTP1) inhibitor. It activates JNK by inhibiting GSTP1 and has shown potential for treating myelodysplastic syndrome (MDS). This compound stimulates both lymphocyte production and bone marrow progenitor proliferation.
  • Activates JNK by inhibiting GSTP1
  • Stimulates lymphocyte production
  • Stimulates bone marrow progenitor proliferation
  • Potential for treating myelodysplastic syndrome (MDS)
Catalog No. 50-003-51163