Learn More
Foxx Life Sciences EZBio Vactrap S Vacuum Trap System (Vacuum Flask Alternative) for Aspiration & Vacuum Protection, Shatterproof 1L Aspirator Bottle Assembly with Secondary Container, Bleach Compatible HDPE, Inline Vent Filter, Sterile, 1/EA
Supplier:  Foxx Life Sciences 6421001FLS
Encompass_Preferred
EZBio Vactrap S is a sterile vacuum trap system for cleanroom bioprocessing, protecting vacuum lines, pumps, and personnel from hazardous liquids and biohazards. It safely contains waste from supernatant removal, chemical separation, and cell culture media aspiration. One bottle collects contaminated fluids for decontamination, while a second serves as overflow protection. A hydrophobic inline filter prevents aerosol spread.
Features
Pre-assembled, sterile, and double-bagged for cleanroom use
Secondary containment prevents spills and overflows
Shatterproof for enhanced safety
Wide-mouth design for easy cleaning
Tube holder prevents spills when not in use
EZBio HP vent filter with 0.2 µm PTFE membrane blocks bacterial aerosols
40 CFR 264.175 compliant
Applications
Cell culture media aspiration
Supernatant removal
Chemical separation in biopharma
Waste collection in high-purity processes
Safe biohazardous waste handling
Catalog No. 50-298-5667