Learn More
Bellco Glass, Inc. EZFlow High Particulate Syringe Filter, 0.22µm PES, Glass Fiber Prefilter, 25mm, 100/PK
SDP
Supplier:  Bellco Glass, Inc. 452800529
Encompass_Preferred
EZFlow 25mm Syringe Filter, .2µm PES,Glass Fiber Prefilter, 100/packEZFlow 25mm high particulate Syringe filters are designed for high throughput filtration of viscous and particle-laden solutions. The syringe filter contains a filter membrane along with a glass fiber pre-filter. The filter membrane is available in PES or Hydrophilic PVDF materials, offered in 0.2µm or 0.45µm pore sizes. The filter membrane is reinforced with a polypropylene housing with a female Luer-Lok inlet and a male Luer Slip outlet.All material comply with USP Class VI Standards.USP Class VISpecificationsProduct Diameter: 1.17" (29.7mm)Product Height: 0.88" (22.4mm)Filter Diameter: 25mmFilter Material: PES w/ Glass Fiber PrefilterFilter Pore Size: .2µm
Catalog No. 50-255-0233