Learn More
Medchemexpress LLC Ezh2-in-13 | 1403255-41-3 | 99.7% | 571.75 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14723010MM/1ML
Encompass_Preferred
EZH2-IN-13 is a potent EZH2 inhibitor, referenced as compound 73 in patent WO2017139404. It can be used for studying cancers or precancerous lesions associated with EZH2 activity. Identified uses include laboratory chemicals and the manufacture of substances.
  • Potent EZH2 inhibitor
  • For research use only
  • Available in solid form and in solution (10 mM in DMSO)
  • Intended for research use by experienced personnel in appropriately equipped and authorized facilities
  • Not fully validated for medical applications
Catalog No. 50-003-61974